  • Search Results
  • Modulation of microenvironmental pH and utilization of alkalizers in crystalline solid dispersion for enhanced solubility and stability of clarithromicin.
Modulation of microenvironmental pH and utilization of alkalizers in crystalline solid dispersion for enhanced solubility and stability of clarithromicin.
Archives of pharmacal research (2014-09-13)
Jun-Bom Park, Young-Joon Park, Chin-Yang Kang, Beom-Jin Lee
ABSTRACT
Clarithromycin (CAM) is known to be poorly water-soluble and acid-labile drug. Various alkalizers such as MgO, Na2CO3, Na2HPO4 and NaHCO3 were utilized to modulate the microenvironmental pH (pHM) and to improve the low stability and solubility of CAM in a crystalline-solid dispersion system (CSD). Polyvinylpyrrolidone (PVP K-30) and hydroxypropylmethylcellulose (HPMC) 4000-based CSDs containing alkalizers were prepared by cosolvent precipitation followed by evaporation process. The dried-CSDs mixed with microcrystalline cellulose, 2% croscarmellose sodium, and 1% magnesium stearate was then directly compressed into tablet. A dissolution test was carried out in 900 mL of pH 5.0 buffer solutions at 37 °C with a 50 rpm paddle speed. pHM, surface morphology, and structural behaviors were investigated. The dissolution rates of CAM in CSD containing alkalizers were improved. The drug in CSD remained crystalline as observed by differential scanning calorimetry and powder X-ray diffraction. Scanning electron microscopy revealed nearly identical images regardless of the sorts and amounts of carriers. PVP-based CSD tablet without alkalizer showed greater drug release, while HPMC-based CSD tablet without alkalizer retarded drug release due to its greater swelling capability. However, when the alkalizers were added in CSD tablet, the drug release was sharply increased. NaHCO3 induced the most rapid drug release while MgO retarded drug dissolution. Alkalizers in CSD also could maintain the pHM of the tablet above pH 5 under acidic conditions. The use of pH modifiers in CSDs could provide a useful method to improve the dissolution rate and stability of CAM via modulation of pHM without changing drug crystallinity.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C