  • Search Results
  • Gum arabic-curcumin conjugate micelles with enhanced loading for curcumin delivery to hepatocarcinoma cells.
Gum arabic-curcumin conjugate micelles with enhanced loading for curcumin delivery to hepatocarcinoma cells.
Carbohydrate polymers (2015-10-03)
P R Sarika, Nirmala Rachel James, P R Anil Kumar, Deepa K Raj, T V Kumary
ABSTRACT
Curcumin is conjugated to gum arabic, a highly water soluble polysaccharide to enhance the solubility and stability of curcumin. Conjugation of curcumin to gum arabic is confirmed by (1)H NMR, fluorescence and UV spectroscopy studies. The conjugate self assembles to spherical nano-micelles (270 ± 5 nm) spontaneously, when dispersed in aqueous medium. Spherical morphology of the self assembled conjugate is evidenced by field emission scanning electron microscopy and transmission electron microscopy. The self assembly of the amphiphilic conjugate into micelle in aqueous medium significantly enhances the solubility (900 fold of that of free curcumin) and stability of curcumin in physiological pH. The anticancer activity of the conjugate micelles is found to be higher in human hepatocellular carcinoma (HepG2) cells than in human breast carcinoma (MCF-7) cells. The conjugate exhibits enhanced accumulation and toxicity in HepG2 cells due to the targeting efficiency of the galactose groups present in gum arabic.
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis