  • Search Results
  • Identification of six new Alternaria sulfoconjugated metabolites by high-resolution neutral loss filtering.
Identification of six new Alternaria sulfoconjugated metabolites by high-resolution neutral loss filtering.
Rapid communications in mass spectrometry : RCM (2015-09-04)
Megan J Kelman, Justin B Renaud, Keith A Seifert, Jonathan Mack, Kumaran Sivagnanam, Ken K-C Yeung, Mark W Sumarah
ABSTRACT
Many species of Alternaria damage important agricultural crops, including small grains and tomatoes. These fungi can produce a variety of secondary metabolites, some of which are toxic to humans and animals. Interest in screening for conjugated or 'modified' mycotoxins has increased because of their tendency to evade traditional analytical screening methods. Two sulfoconjugated Alternaria toxins have been reported and the potential exists for many more. One hundred and forty-eight Canadian strains of Alternaria spp., about half of them isolated from grain, were grown on Potato Dextrose Agar in Petri dishes for 7 days. Plugs of each strain were removed, extracted and screened by a rapid liquid chromatography (LC)/data-dependent tandem mass spectrometry (MS(2)) method in negative electrospray ionization mode. Data generated on an Orbitrap Q-Exactive mass spectrometer was processed by post-acquisition neutral loss filtering (NLF). Seven isolates that produced sulfoconjugates of known Alternaria toxins were selected for growth on three additional types of fermentation media. Collision-induced dissociation of sulfoconjugated ions displayed a distinctive neutral loss of SO3 (79.957 Da) that was detected in the MS(2) datasets using post-acquisition NLF. A total of 108 of the 148 isolates screened produced sulfoconjugated metabolites on agar plates. Analysis of the seven isolates grown in liquid culture, on rice and Cheerios, led to the discovery of six new, two previously reported and 30 unidentified sulfoconjugated compounds. NLF of HRMS(2) data from an Orbitrap Q-Exactive is a powerful tool for the rapid discovery of sulfoconjugated fungal metabolites. This technique could also be applied to the detection of other important conjugated mycotoxins such as glucosides. The majority of the Canadian isolates of Alternaria spp. studied produced sulfoconjugated metabolites, some of which had no known 'free' Alternaria precursor metabolite, indicating that they are possibly new metabolites. The advantage of sulfoconjugation to Alternaria spp. is unknown, and warrants further study into the mechanisms behind the sulfur assimilatory pathways.
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles