Supervised discriminant analysis for droplet micro-magnetofluidics.
Microfluidics and nanofluidics (2015-09-18)
Gungun Lin, Vladimir M Fomin, Denys Makarov, Oliver G Schmidt
ABSTRACT
We apply the technique of supervised discriminant analysis (SDA) for in-flow detection in droplet-based magnetofluidics. Based on the SDA, we successfully discriminate
Sigma-Aldrich
Mineral oil, light oil