  • Search Results
  • Evaluation of rhamnolipid (RL) as a biosurfactant for the removal of chromium from aqueous solutions by precipitate flotation.
Evaluation of rhamnolipid (RL) as a biosurfactant for the removal of chromium from aqueous solutions by precipitate flotation.
Journal of environmental management (2015-10-05)
Ali Salmani Abyaneh, Mohammad Hassan Fazaelipoor
ABSTRACT
Rhamnolipid (RL) is a biosurfactant which is produced by the bacterial species Pseudomonas aeruginosa. Although applications of this material have been examined in various fields, its applications in the flotation of heavy metals remain to be explored. In this research, rhamnolipid was applied as a collector in the flotation of Cr(III) from aqueous solutions. FeSO4 was used for the precipitation of Cr(VI) to Cr(III) which was subsequently removed by flotation. A two level full factorial design (with center points) was used to evaluate the effects of pH, air flow rate, RL/Cr and Fe/Cr molar ratios on the performance of the flotation system. The results showed that the biosurfactant was highly effective in the removal of chromium, and all of the factors had significant effects on the flotation performance. The chromium removal efficiencies of greater than 95% were obtained with the initial chromium concentration of 40 ppm within 5 min. Kinetic studies showed that a first order kinetic model was appropriate to describe the precipitate flotation of Cr(III) using rhamnolipid as a collector. The interference of NaCl, CaCl2, CaSO4, and CaCO3 on the Cr removal was also investigated, and it was demonstrated that CaSO4 and CaCO3 as sparingly water soluble salts, and CaCl2 as a contributor to water hardness had significant negative impacts on Cr removal efficiency of rhamnolipid.
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%