  • Search Results
  • Hydroxyproline metabolism in a mouse model of Primary Hyperoxaluria Type 3.
Hydroxyproline metabolism in a mouse model of Primary Hyperoxaluria Type 3.
Biochimica et biophysica acta (2015-10-03)
Xingsheng Li, John Knight, W Todd Lowther, Ross P Holmes
ABSTRACT
Primary Hyperoxaluria Type 3 is a recently discovered form of this autosomal recessive disease that results from mutations in the gene coding for 4-hydroxy-2-oxoglutarate aldolase (HOGA1). This enzyme is one of the 2 unique enzymes in the hydroxyproline catabolism pathway. Affected individuals have increased urinary excretions of oxalate, 4-hydroxy-L-glutamate (4-OH-Glu), 4-hydroxy-2-oxoglutarate (HOG), and 2,4-dihydroxyglutarate (DHG). While 4-OH-Glu and HOG are precursor substrates of HOGA1 and increases in their concentrations are expected, how DHG is formed and how HOG to oxalate are unclear. To resolve these important questions and to provide insight into possible therapeutic avenues for treating this disease, an animal model of the disease would be invaluable. We have developed a mouse model of this disease which has null mutations in the Hoga1 gene and have characterized its phenotype. It shares many characteristics of the human disease, particularly when challenged by the inclusion of hydroxyproline in the diet. An increased oxalate excretion is not observed in the KO mice which may be consistent with the recent recognition that only a small fraction of the individuals with the genotype for HOGA deficiency develop PH.
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis