  • Search Results
  • Liver injury-on-a-chip: microfluidic co-cultures with integrated biosensors for monitoring liver cell signaling during injury.
Liver injury-on-a-chip: microfluidic co-cultures with integrated biosensors for monitoring liver cell signaling during injury.
Lab on a chip (2015-10-20)
Qing Zhou, Dipali Patel, Timothy Kwa, Amranul Haque, Zimple Matharu, Gulnaz Stybayeva, Yandong Gao, Anna Mae Diehl, Alexander Revzin
ABSTRACT
Tissue injury triggers complex communication between cells via secreted signaling molecules such as cytokines and growth factors. Discerning when and where these signals begin and how they propagate over time is very challenging with existing cell culture and analysis tools. The goal of this study was to develop new tools in the form of microfluidic co-cultures with integrated biosensors for local and continuous monitoring of secreted signals. Specifically, we focused on how alcohol injury affects TGF-β signaling between two liver cell types, hepatocytes and stellate cells. Activation of stellate cells happens early during liver injury and is at the center of liver fibrosis. We demonstrated that alcohol injury to microfluidic co-cultures caused significantly higher levels of stellate cell activation compared to conditioned media and transwell injury experiments. This highlighted the advantage of the microfluidic co-culture: placement of two cell types in close proximity to ensure high local concentrations of injury-promoting secreted signals. Next, we developed a microsystem consisting of five chambers, two for co-culturing hepatocytes with stellate cells and three additional chambers containing miniature aptamer-modified electrodes for monitoring secreted TGF-β. Importantly, the walls separating microfluidic chambers were actuatable; they could be raised or lowered to create different configurations of the device. The use of reconfigurable microfluidics and miniature biosensors revealed that alcohol injury causes hepatocytes to secrete TGF-β molecules, which diffuse over to neighboring stellate cells and trigger production of additional TGF-β from stellate cells. Our results lend credence to the emerging view of hepatocytes as active participants of liver injury. Broadly speaking, our microsystem makes it possible to monitor paracrine crosstalk between two cell types communicating via the same signaling molecule (e.g. TGF-β).
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Methylene blue, certified by the Biological Stain Commission
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
SAFC
HEPES
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.