  • Search Results
  • Bone Morphogenic Protein 4 Mediates NOX1-Dependent eNOS Uncoupling, Endothelial Dysfunction, and COX2 Induction in Type 2 Diabetes Mellitus.
Bone Morphogenic Protein 4 Mediates NOX1-Dependent eNOS Uncoupling, Endothelial Dysfunction, and COX2 Induction in Type 2 Diabetes Mellitus.
Molecular endocrinology (Baltimore, Md.) (2015-06-30)
Ji-Youn Youn, Jun Zhou, Hua Cai
ABSTRACT
We have recently shown that angiotensin II-mediated uncoupling of endothelial nitric oxide synthase (eNOS) contributes to endothelial dysfunction in streptozotocin-induced type 1 diabetes mellitus. However, it has remained unclear whether and how eNOS uncoupling occurs in type 2 diabetes mellitus (T2DM) and the consequences of such in regulating vascular function. Here we investigated a role of bone morphogenic protein (BMP)-4 in mediating eNOS uncoupling, endothelial dysfunction, and inflammation in db/db mice. Circulating levels of BMP4 were markedly elevated in db/db mice but not in mice with type 1 diabetes mellitus, in which angiotensin II levels were significantly increased. Infusion of BMP4 antagonist noggin into db/db mice (15 μg/kg/day, 4 weeks) abolished eNOS uncoupling activity while restoring tetrahydrobiopterin (H(4)B) bioavailability. The impaired endothelium-dependent vasorelaxation in db/db aortas was significantly improved by noggin infusion. Exposure of aortic endothelial cells to BMP4 (50 ng/mL, 24 hours) resulted in eNOS uncoupling, which was attenuated by H(4)B precursor sepiapterin or small interfering RNA silencing nicotinamide adenine dinucleotide phosphate oxidase isoform 1 (NOX1). Interestingly, BMP4-dependent NOX1 up-regulation was abrogated by sepiapterin, implicating a NOX1-uncoupled eNOS-NOX1 feed-forward loop. BMP4 induction of cyclooxygenase 2 (COX2) expression and vascular cell adhesion protein 1 was found in db/db mice. Consistently, COX2 was up-regulated by BMP4 in endothelial cells, which was attenuated by sepiapterin, implicating an upstream role of eNOS uncoupling in COX2-mediated inflammatory activation. Taken together, our data for the first time reveal a novel role of BMP4 in inducing NOX1-dependent eNOS uncoupling in T2DM, which may promote development of novel therapeutics restoring endothelial function in T2DM.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture