  • Search Results
  • Activating Mutations in PIK3CA Lead to Widespread Modulation of the Tyrosine Phosphoproteome.
Activating Mutations in PIK3CA Lead to Widespread Modulation of the Tyrosine Phosphoproteome.
Journal of proteome research (2015-08-13)
Muhammad Saddiq Zahari, Xinyan Wu, Brian G Blair, Sneha M Pinto, Raja S Nirujogi, Christine A Jelinek, Radhika Malhotra, Min-Sik Kim, Ben Ho Park, Akhilesh Pandey
ABSTRACT
The human oncogene PIK3CA is frequently mutated in human cancers. Two hotspot mutations in PIK3CA, E545K and H1047R, have been shown to regulate widespread signaling events downstream of AKT, leading to increased cell proliferation, growth, survival, and motility. We used quantitative mass spectrometry to profile the global phosphotyrosine proteome of isogenic knock-in cell lines containing these activating mutations, where we identified 824 unique phosphopeptides. Although it is well understood that these mutations result in hyperactivation of the serine/threonine kinase AKT, we found a surprisingly widespread modulation of tyrosine phosphorylation levels of proteins in the mutant cells. In the tyrosine kinome alone, 29 tyrosine kinases were altered in their phosphorylation status. Many of the regulated phosphosites that we identified were located in the kinase domain or the canonical activation sites, indicating that these kinases and their downstream signaling pathways were activated. Our study demonstrates that there is frequent and unexpected cross-talk that occurs between tyrosine signaling pathways and serine/threonine signaling pathways activated by the canonical PI3K-AKT axis.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
SAFC
L-Arginine
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
DL-Arginine, ≥95% (NT)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
L-Glutamine