MilliporeSigma
  • Two short sequences in OsNAR2.1 promoter are necessary for fully activating the nitrate induced gene expression in rice roots.
Two short sequences in OsNAR2.1 promoter are necessary for fully activating the nitrate induced gene expression in rice roots.
Scientific reports (2015-07-08)
Xiaoqin Liu, Huimin Feng, Daimin Huang, Miaoquan Song, Xiaorong Fan, Guohua Xu
ABSTRACT
Nitrate is an essential nitrogen source and serves as a signal to control growth and gene expression in plants. In rice, OsNAR2.1 is an essential partner of multiple OsNRT2 nitrate transporters for nitrate uptake over low and high concentration range. Previously, we have reported that -311 bp upstream fragment from the translational start site in the promoter of OsNAR2.1 gene is the nitrate responsive region. To identify the cis-acting DNA elements necessary for nitrate induced gene expression, we detected the expression of beta-glucuronidase (GUS) reporter in the transgenic rice driven by the OsNAR2.1 promoter with different lengths and site mutations of the 311 bp region. We found that -129 to -1 bp region is necessary for the nitrate-induced full activation of OsNAR2.1. Besides, the site mutations showed that the 20 bp fragment between -191 and -172 bp contains an enhancer binding site necessary to fully drive the OsNAR2.1 expression. Part of the 20 bp fragment is commonly presented in the sequences of different promoters of both the nitrate induced NAR2 genes and nitrite reductase NIR1 genes from various higher plants. These findings thus reveal the presence of conserved cis-acting element for mediating nitrate responses in plants.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C