  • Search Results
  • Alveolar Epithelial A2B Adenosine Receptors in Pulmonary Protection during Acute Lung Injury.
Alveolar Epithelial A2B Adenosine Receptors in Pulmonary Protection during Acute Lung Injury.
Journal of immunology (Baltimore, Md. : 1950) (2015-07-19)
Sandra Hoegl, Kelley S Brodsky, Michael R Blackburn, Harry Karmouty-Quintana, Bernhard Zwissler, Holger K Eltzschig
ABSTRACT
Acute lung injury (ALI) is an acute inflammatory lung disease that causes morbidity and mortality in critically ill patients. However, there are many instances where ALI resolves spontaneously through endogenous pathways that help to control excessive lung inflammation. Previous studies have implicated the extracellular signaling molecule adenosine and signaling events through the A2B adenosine receptor in lung protection. In this context, we hypothesized that tissue-specific expression of the A2B adenosine receptor is responsible for the previously described attenuation of ALI. To address this hypothesis, we exposed mice with tissue-specific deletion of Adora2b to ALI, utilizing a two-hit model where intratracheal LPS treatment is followed by injurious mechanical ventilation. Interestingly, a head-to-head comparison of mice with deletion of Adora2b in the myeloid lineage (Adora2b(loxP/loxP) LysM Cre(+)), endothelial cells (Adora2b(loxP/loxP) VE-cadherin Cre(+)), or alveolar epithelial cells (Adora2b(loxP/loxP) SPC Cre(+)) revealed a selective increase in disease susceptibility in Adora2b(loxP/loxP) SPC Cre(+) mice. More detailed analysis of Adora2b(loxP/loxP) SPC Cre(+) mice confirmed elevated lung inflammation and attenuated alveolar fluid clearance. To directly deliver an A2B adenosine receptor-specific agonist to alveolar epithelial cells, we subsequently performed studies with inhaled BAY 60-6583. Indeed, aerosolized BAY 60-6583 treatment was associated with attenuated pulmonary edema, improved histologic lung injury, and dampened lung inflammation. Collectively, these findings suggest that alveolar epithelial A2B adenosine receptor signaling contributes to lung protection, and they implicate inhaled A2B adenosine receptor agonists in ALI treatment.
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M