  • Search Results
  • Isoflavone pharmacokinetics and metabolism after consumption of a standardized soy and soy-almond bread in men with asymptomatic prostate cancer.
Isoflavone pharmacokinetics and metabolism after consumption of a standardized soy and soy-almond bread in men with asymptomatic prostate cancer.
Cancer prevention research (Philadelphia, Pa.) (2015-08-16)
Jennifer H Ahn-Jarvis, Steven K Clinton, Elizabeth M Grainger, Kenneth M Riedl, Steven J Schwartz, Mei-Ling T Lee, Raul Cruz-Cano, Gregory S Young, Gregory B Lesinski, Yael Vodovotz
ABSTRACT
Epidemiologic associations suggest that populations consuming substantial amounts of dietary soy exhibit a lower risk of prostate cancer. A 20-week randomized, phase II, crossover trial was conducted in 32 men with asymptomatic prostate cancer. The crossover involved 8 weeks each of soy bread (SB) and soy-almond bread (SAB). The primary objective was to investigate isoflavone bioavailability and metabolite profile. Secondary objectives include safety, compliance, and assessment of biomarkers linked to prostate carcinogenesis. Two distinct SBs were formulated to deliver approximately 60 mg aglycone equivalents of isoflavones per day. The isoflavones were present as aglycones (∼78% as aglycones) in the SAB whereas in the standard SB predominantly as glucosides (18% total isoflavones as aglycones). Compliance to SB (97% ± 4%) and SAB (92% ± 18%) was excellent; toxicity was rare and limited to grade 1 gastrointestinal complaints. Pharmacokinetic studies between SB and SAB showed modest differences. Peak serum concentration time (Tmax) was significantly faster with SAB meal compared with SB in some isoflavonoids, and AUC0 to 24 h of dihydrodaidzein and O-desmethylangolensin was significantly greater after an SB meal. An exploratory cluster analysis was used to identify four isoflavone-metabolizing phenotypes. Insulin-like growth factor-binding protein increased significantly by 41% (P = 0.024) with soy intervention. Findings from this study provide the necessary framework to study isoflavone-metabolizing phenotypes as a strategy for identification of individuals that might benefit or show resistance to cancer preventive strategies using dietary soy. A standardized SB used for future large-scale randomized clinical trials to affect human prostate carcinogenesis is feasible.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Glycitein, ≥97% (HPLC)
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Daidzin, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid