  • Search Results
  • Green Tea Lowers Hepatic COX-2 and Prostaglandin E2 in Rats with Dietary Fat-Induced Nonalcoholic Steatohepatitis.
Green Tea Lowers Hepatic COX-2 and Prostaglandin E2 in Rats with Dietary Fat-Induced Nonalcoholic Steatohepatitis.
Journal of medicinal food (2014-12-03)
Min-Yu Chung, Eunice Mah, Christopher Masterjohn, Sang K Noh, Hea Jin Park, Richard M Clark, Young-Ki Park, Ji-Young Lee, Richard S Bruno
ABSTRACT
Green tea extract (GTE) protects against nonalcoholic steatohepatitis (NASH) by decreasing hepatic steatosis and nuclear factor kappa B (NFκB) activation. We hypothesized that hypolipidemic and anti-inflammatory activities of GTE would protect against NASH by reducing cyclooxygenase-2 (COX-2), an NFκB-dependent enzyme, and prostaglandin E2 (PGE2) in a dietary fat-induced obese model. Male Wistar rats were fed a low-fat diet containing no GTE or a high-fat (HF) diet containing GTE at 0%, 1%, or 2% for 8 weeks. Insulin resistance and total hepatic fatty acids increased following HF feeding (P<.05) and these were normalized by GTE at 1-2%. GTE (1-2%) normalized hepatic malondialdehyde without affecting cytochrome P450 2E1 mRNA expression, which was otherwise increased by HF feeding. HF-mediated increases in hepatic COX-2 protein and activity as well as PGE2 concentrations were normalized by GTE (1-2%). COX-2 activity and PGE2 were correlated to each other, and to serum alanine aminotransferase (ALT) and hepatic NFκB-binding activity (P<.05; r=0.28-0.49). GTE attenuated HF-mediated increases in total hepatic n-6 and n-3, without affecting the n-6/n-3 ratio. GTE did not affect HF-mediated increases in n-6 in nonesterified fatty acid (NEFA) and phospholipid pools, whereas n-3 and n-6/n-3 in both pools were unaffected by GTE and HF feeding. GTE decreased total hepatic arachidonic acid without affecting HF-mediated increases in arachidonic acid in NEFA or phospholipid pools. Thus, GTE attenuates lipid peroxidation and PGE2 accumulation by decreasing COX-2 activity independent of arachidonic acid availability and supports an additional mechanism by which GTE protects against liver injury during NASH in an HF-feeding model.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
SAFC
HEPES
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture