  • Search Results
  • Anti-inflammatory activity and phenolic profile of propolis from two locations in Región Metropolitana de Santiago, Chile.
Anti-inflammatory activity and phenolic profile of propolis from two locations in Región Metropolitana de Santiago, Chile.
Journal of ethnopharmacology (2015-04-04)
Gabriela Valenzuela-Barra, Consuelo Castro, Catalina Figueroa, Andrés Barriga, Ximena Silva, Beatriz de Las Heras, Sonsoles Hortelano, Carla Delporte
ABSTRACT
Propolis has long been used as a popular folk medicine due to its wide spectrum of alleged biological and pharmaceutical properties. In Chile, propolis is widely used by folklore medicine as an anti-inflammatory agent; however, this property has not been demonstrated by scientific methods. The objective of this study was to determine the anti-inflammatory activity in vivo and in vitro and to establish the phenolic profile of propolis collected in two localities in Región Metropolitana de Santiago (RM), Chile. Propolis was collected in the areas of Caleu and Buin, RM Chile. Following that, the samples were unwaxed to obtain the global ethanolic extracts of propolis (EEPs) and, from these, the serial extracts of dichloromethane (EEP-DCMs) and ethanol (EEP-EtOHs). The topic anti-inflammatory effect was evaluated through mice ear edema induced by arachidonic acid (AA) and 12-O-tetradecanoylphorbol-13-acetate (TPA) at a dose of 3 mg/ear. Nitric oxide (NO) measurements were determined spectrophotometrically (Greiss reagent) by the accumulation of nitrite in the medium of macrophages RAW 264.7 stimulated with the lipopolysaccharide (LPS, 1 μg/mL) for 20 h at different concentrations of the EEPs, EEP-DCMs and EEP-EtOHs (6.25-50.00 μg/mL). The content of total phenols and flavonoids were determined through the methods of Folin-Ciocalteau and AlCl3, respectively. The profile of phenolic compounds was determined by HPLC-UV-ESI-MS/MS. The EEP-EtOH (64%) and EEP (59%) of Buin were the most active in the inflammation induced by TPA and AA respectively, being the anti-inflammatory effect stronger than the same Caleu extracts. Regarding the release of NO, all the extracts from the Buin propolis inhibited significantly its release in a concentration-dependent manner, this inhibition was stronger than the extracts from Caleu propolis. Our research shows for the first time a comparative study of the topical in vivo activity of two Chilean propolis. Both propolis showed in vivo topical anti-inflammatory activity against AA and TPA, the most active was Buin propolis and this difference is due in part to the variations in total phenols and flavonoids content and the phenolic profile. The phenols and flavonoids content of Buin propolis was higher than Caleu propolis. The extracts from Buin propolis result in a lower release of NO.
Sigma-Aldrich
Phenol, natural, 97%, FG
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)