  • Search Results
  • Dual blockade of PI3K/AKT/mTOR (NVP-BEZ235) and Ras/Raf/MEK (AZD6244) pathways synergistically inhibit growth of primary endometrioid endometrial carcinoma cultures, whereas NVP-BEZ235 reduces tumor growth in the corresponding xenograft models.
Dual blockade of PI3K/AKT/mTOR (NVP-BEZ235) and Ras/Raf/MEK (AZD6244) pathways synergistically inhibit growth of primary endometrioid endometrial carcinoma cultures, whereas NVP-BEZ235 reduces tumor growth in the corresponding xenograft models.
Gynecologic oncology (2015-05-03)
Stefanie Schrauwen, Jeroen Depreeuw, Lieve Coenegrachts, Els Hermans, Diether Lambrechts, Frédéric Amant
ABSTRACT
Endometrial carcinoma (EC) is the most common gynecological cancer in the Western World. Treatment options are limited for advanced and recurrent disease. Therefore, new treatment options are necessary. Inhibition of the PI3K/AKT/mTOR and/or the Ras/Raf/MEK pathways is suggested to be clinically relevant. However, the knowledge about the effect of combination targeted therapy in EC is limited. The aim of this study was to investigate the effect of these therapies on primary endometrioid EC cell cultures in vitro and in vivo. Primary endometrioid EC cell cultures were incubated with Temsirolimus (mTORC1 inhibitor), NVP-BKM120 (pan-PI3K inhibitor), NVP-BEZ235 (pan-PI3K/mTOR inhibitor), or AZD6244 (MEK1/2 inhibitor) as single treatment. In vitro, the effect of NVP-BEZ235 with or without AZD6244 was determined for cell viability, cell cycle arrest, apoptosis induction, and cell signaling. In vivo, the effect of NVP-BEZ35 was investigated for 2 subcutaneous xenograft models of the corresponding primary cultures. NVP-BEZ235 was the most potent PI3K/AKT/mTOR pathway inhibitor. NVP-BEZ235 and AZD6244 reduced cell viability and induced cell cycle arrest and apoptosis, by reduction of p-AKT, p-S6, and p-ERK levels. Combination treatment showed a synergistic effect. In vivo, NVP-BEZ235 reduced tumor growth and inhibited p-S6 expression. The effects of the compounds were independent of the mutation profile of the cell cultures used. A synergistic antitumor effect was shown for NVP-BEZ235 and AZD6244 in primary endometrioid EC cells in vitro. In addition, NVP-BEZ235 induced reduction of tumor growth in vivo. Therefore, targeted therapies seem an interesting strategy to further evaluate in clinical trials.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
SAFC
L-Glutamine
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Temsirolimus, ≥98% (HPLC)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Mammalian Cell Lysis Kit
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)