  • Search Results
  • Some South African Rubiaceae Tree Leaf Extracts Have Antimycobacterial Activity Against Pathogenic and Non-pathogenic Mycobacterium Species.
Some South African Rubiaceae Tree Leaf Extracts Have Antimycobacterial Activity Against Pathogenic and Non-pathogenic Mycobacterium Species.
Phytotherapy research : PTR (2015-04-11)
Abimbola O Aro, Jean P Dzoyem, Tiny M Hlokwe, Evelyn Madoroba, Jacobus N Eloff, Lyndy J McGaw
ABSTRACT
Tuberculosis (TB) caused by Mycobacterium tuberculosis remains an ongoing threat to human health. Many plant species contain antimycobacterial compounds, which may serve as template molecules for new anti-TB drugs. The Rubiaceae family is the largest family of trees in southern Africa, and preliminary evidence revealed antimycobacterial activity in several species of the genus, motivating further studies. Leaf extracts of 15 tree species from the Rubiaceae family were screened for antimycobacterial activity against pathogenic M. tuberculosis and non-pathogenic Mycobacterium smegmatis, Mycobacterium aurum and Mycobacterium bovis BCG (Bacillus Calmette-Guérin) using a twofold serial microdilution assay. Cytotoxicity was determined using a tetrazolium-based colorimetric assay against C3A liver cells and Vero kidney cells. Minimum inhibitory concentration values as low as 0.04 mg/mL against M. smegmatis and M. tuberculosis were recorded. Activity against M. aurum was the best predictor of activity against pathogenic M. tuberculosis (correlation coefficient = 0.9). Bioautography indicated at least 40 different antimycobacterial compounds in the extracts. Cytotoxicity of the extracts varied, and Oxyanthus speciosus had the most promising selectivity index values.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
MTT Formazan, powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
MTT Formazan, MTT reduction end product
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC