  • Search Results
  • Characterization and simultaneous quantification of biological aporphine alkaloids in Litsea cubeba by HPLC with hybrid ion trap time-of-flight mass spectrometry and HPLC with diode array detection.
Characterization and simultaneous quantification of biological aporphine alkaloids in Litsea cubeba by HPLC with hybrid ion trap time-of-flight mass spectrometry and HPLC with diode array detection.
Journal of separation science (2015-05-27)
Shuiying Zhang, Qian Zhang, Qiang Guo, Yunfang Zhao, Xiaoli Gao, Xingyun Chai, Pengfei Tu
ABSTRACT
The root and rhizome of Litsea cubeba (Lour) Pers., named 'Dou-chi-jiang' in Chinese, has been traditionally used for treatment of cardiovascular and cerebrovascular diseases, rheumatic arthralgia, and other diseases in China. Aporphine alkaloids are its characteristic ingredients and responsible for its bioactivities, especially anti-inflammatory and analgesic effects. A sensitive and reliable high-performance liquid chromatography with diode array detection-tandem mass spectrometry method was developed for characterization and simultaneous determination of biological aporphine alkaloids in 'Dou-chi-jiang'. The optimized chromatographic conditions were performed on an Eclipse XDB C18 column with a gradient of acetonitrile/water containing 0.1% formic acid as the mass spectrometry mobile phase and acetonitrile/water containing 0.2% diethylamine (pH 3.10, adjusted by acetic acid) as the liquid chromatography mobile phase. The fragmentation pathways by loss of CO, ·CH3 , ·NH3 , and ·NH2 CH3 were detected as characteristic for aporphine alkaloids. Based on these characteristics, total 12 analogues were identified. The quantification method was validated in terms of linearity, precision, and accuracy for six major aporphine alkaloids, which was successfully applied for simultaneous determination in ten batches of samples. The established method is simple, rapid, and specific for characterization and quantitation of aporphine alkaloids in 'Dou-chi-jiang' and other traditional Chinese medicines rich in this kind of ingredient.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Diethylamine, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Diethylamine, puriss. p.a., ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Pricing and availability is not currently available.
SAFC
Acetic acid, glacial, PharmaGrade, USP, JP, Ph. Eur., Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
Pricing and availability is not currently available.
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Diethylamine, purified by redistillation, 99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Diethylamine, BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Pricing and availability is not currently available.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, NMR reference standard
Pricing and availability is not currently available.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, NMR reference standard, 0.23 wt. % in D2O (99.9 atom % D), water 0.05 wt. %, NMR tube size 6.5 mm × 8 in.
Pricing and availability is not currently available.
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Pricing and availability is not currently available.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.