  • Search Results
  • The WRKY45-Dependent Signaling Pathway Is Required For Resistance against Striga hermonthica Parasitism.
The WRKY45-Dependent Signaling Pathway Is Required For Resistance against Striga hermonthica Parasitism.
Plant physiology (2015-05-31)
J Musembi Mutuku, Satoko Yoshida, Takafumi Shimizu, Yasunori Ichihashi, Takanori Wakatake, Akira Takahashi, Mitsunori Seo, Ken Shirasu
ABSTRACT
The root hemiparasite witchweed (Striga spp.) is a devastating agricultural pest that causes losses of up to $1 billion US annually in sub-Saharan Africa. Development of resistant crops is one of the cost-effective ways to address this problem. However, the molecular mechanisms underlying resistance are not well understood. To understand molecular events upon Striga spp. infection, we conducted genome-scale RNA sequencing expression analysis using Striga hermonthica-infected rice (Oryza sativa) roots. We found that transcripts grouped under the Gene Ontology term defense response were significantly enriched in up-regulated differentially expressed genes. In particular, we found that both jasmonic acid (JA) and salicylic acid (SA) pathways were induced, but the induction of the JA pathway preceded that of the SA pathway. Foliar application of JA resulted in higher resistance. The hebiba mutant plants, which lack the JA biosynthesis gene allene oxide cyclase, exhibited severe S. hermonthica susceptibility. The resistant phenotype was recovered by application of JA. By contrast, the SA-deficient NahG rice plants were resistant against S. hermonthica, indicating that endogenous SA is not required for resistance. However, knocking down WRKY45, a regulator of the SA/benzothiadiazole pathway, resulted in enhanced susceptibility. Interestingly, NahG plants induced the JA pathway, which was down-regulated in WRKY45-knockdown plants, linking the resistant and susceptible phenotypes to the JA pathway. Consistently, the susceptibility phenotype in the WRKY45-knockdown plants was recovered by foliar JA application. These results point to a model in which WRKY45 modulates a cross talk in resistance against S. hermonthica by positively regulating both SA/benzothiadiazole and JA pathways.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Phytol, ≥97%, FG
Sigma-Aldrich
Phytol, 97%, mixture of isomers
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Pyrocatechol, suitable for plant cell culture, BioReagent, ≥99%, powder
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile
Supelco
Acetonitrile, HPLC grade, ≥99.93%