  • Search Results
  • A metabolomics approach to studying the effects of Jinxin oral liquid on RSV-infected mice using UPLC/LTQ-Orbitrap mass spectrometry.
A metabolomics approach to studying the effects of Jinxin oral liquid on RSV-infected mice using UPLC/LTQ-Orbitrap mass spectrometry.
Journal of ethnopharmacology (2015-08-04)
Li-na Du, Tong Xie, Jian-ya Xu, An Kang, Liu-qing Di, Jin-jun Shan, Shou-chuan Wang
ABSTRACT
Jinxin oral liquid (JOL) is a traditional Chinese medicine (TCM) formula modified from ma-xing-shi-gan-tang, an ancient formula widely used in the treatment of respiratory diseases such as bronchitis, pneumonia, and asthma. In our previous studies, JOL was shown to safely and effectively treat viral pneumonia, especially that involving respiratory syncytial virus (RSV). To investigate the mechanism of the effect of JOL in RSV infected mice, using a metabolomics approach based on ultra-performance liquid chromatography coupled with linear ion trap quadrupole-Orbitrap mass spectrometry (UPLC/LTQ-Orbitrap-MS). BALB/c mice were divided into four groups, the control group (saline inoculation/no treatment), RSV group (RSV inoculation/saline treatment), RSV+JOL group (RSV inoculation/JOL treatment), and RSV+Riba group (RSV inoculation/ribavirin treatment). Plasma and lung tissue samples were collected 7 days after the inoculation/treatment protocols, and UPLC/LTQ-Orbitrap-MS method based on metabolomics was developed. Principal component analysis (PCA) and orthogonal partial least squares-discriminant analysis (OPLS-DA) were utilized to identify biomarkers potentially associated with the anti-RSV activity of JOL. JOL was associated with reduced inflammatory responses in RSV-infected lung tissue. The combination of PCA and OPLS-DA revealed deviations in 11 biomarkers in plasma, and 16 biomarkers in lung tissue induced by RSV that were corrected with JOL treatment. These biomarkers were primarily components of metabolic pathways involving glycerophosphocholines, sphingolipids, and glycerolipids. JOL was able to restore the abnormal levels of these biomarkers detected in the plasma and lung tissue of RSV-infected mice to approximately normal levels. This study suggested that JOL can treat RSV pneumonia effectively, partially by ameliorating the associated disturbances to lipid metabolism. The results provided insight into the anti-RSV mechanism of JOL, and also demonstrated that metabolomics is a valuable tool for investigating the efficacy of TCM treatment for RSV pneumonia, and the associated biomarkers involved.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Dibutyl ether, anhydrous, 99.3%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%