  • Search Results
  • Transcriptional regulatory network of SOX4 during myoblast differentiation.
Transcriptional regulatory network of SOX4 during myoblast differentiation.
Biochemical and biophysical research communications (2015-05-15)
Sang-Min Jang, Chul-Hong Kim, Jung-Woong Kim, Kyung-Hee Choi
ABSTRACT
The construction of transcriptional regulatory networks of transcription factors (TFs) has become more important and attractive to understand the alterations of binding protein-dependent transcriptional activity that governs the changes in spatiotemporal expression of TF target genes required in various cellular processes. Therefore, identification of new inner modules including target genes and protein interactions involved in unveiled TF-based transcription networks is currently in the research spotlight. In this study, we reveal a possible SOX4-centered transcriptional network by the identification of novel binding partners and target genes of the TF SOX4 using various screening techniques. Lamin B2, barrier to autointegration factor 1, and apolipoprotein C-III were identified as novel interacting partners of SOX4 by yeast two-hybrid screening, and the genes encoding lysosomal-associated membrane protein 1, ubiquitin-conjugating enzyme E2S, and Map2k2 were identified as putative target genes of SOX4. Differently from the computational networks of TFs, we revealed a SOX4-centered physical network during myoblast differentiation. These results will provide opportunities to better understand the SOX4-centered transcriptional regulation network and TF-based specific gene expression in various cellular environments.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri