  • Search Results
  • Development of an Animal Model for Alcoholic Liver Disease in Zebrafish.
Development of an Animal Model for Alcoholic Liver Disease in Zebrafish.
Zebrafish (2015-04-30)
Jiun-Nong Lin, Lin-Li Chang, Chung-Hsu Lai, Kai-Jen Lin, Mei-Fang Lin, Chih-Hui Yang, Hsi-Hsun Lin, Yen-Hsu Chen
ABSTRACT
Alcoholic liver disease (ALD) continues to be a major cause of liver-related morbidity and mortality worldwide. To date, no zebrafish animal model has demonstrated the characteristic manifestations of ALD in the setting of chronic alcohol exposure. The aim of this study was to develop a zebrafish animal model for ALD. Male adult zebrafish were housed in a 1% (v/v) ethanol solution up to 3 months. A histopathological study showed the characteristic features of alcoholic liver steatosis and steatohepatitis in the early stages of alcohol exposure, including fat droplet accumulation, ballooning degeneration of the hepatocytes, and Mallory body formation. As the exposure time increased, collagen deposition in the extracellular matrix was observed by Sirius red staining and immunofluorescence staining. Finally, anaplastic hepatocytes with pleomorphic nuclei were arranged in trabecular patterns and formed nodules in the zebrafish liver. Over the time course of 1% ethanol exposure, upregulations of lipogenesis, fibrosis, and tumor-related genes were also revealed by semiquantitative and quantitative real-time reverse transcription-polymerase chain reaction. As these data reflect characteristic liver damage by alcohol in humans, this zebrafish animal model may serve as a powerful tool to study the pathogenesis and treatment of ALD and its related disorders in humans.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%