  • Search Results
  • Discovery of a selective, safe and novel anti-malarial compound with activity against chloroquine resistant strain of Plasmodium falciparum.
Discovery of a selective, safe and novel anti-malarial compound with activity against chloroquine resistant strain of Plasmodium falciparum.
Scientific reports (2015-09-09)
Ankita Agarwal, Sarvesh Paliwal, Ruchi Mishra, Swapnil Sharma, Anil Kumar Dwivedi, Renu Tripathi, Sarika Gunjan
ABSTRACT
In recent years the DNA minor groove has attracted much attention for the development of anti-malarial agents. In view of this we have attempted to discover novel DNA minor groove binders through in-silico and in-vitro workflow. A rigorously validated pharmacophore model comprising of two positive ionizable (PI), one hydrophobic (HY) and one ring aromatic (RA) features was used to mine NCI chemical compound database. This led to retrieval of many hits which were screened on the basis of estimated activity, fit value and Lipinski's violation. Finally two compounds NSC639017 and NSC371488 were evaluated for their in-vitro anti-malarial activities against Plasmodium falciparum 3D7 (CQ sensitive) and K1 (CQ resistant) strains by SYBR green-I based fluorescence assay. The results revealed that out of two, NSC639017 posses excellent anti-malarial activity particularly against chloroquine resistant strain and moreover NSC639017 also appeared to be safe (CC50 126.04 μg/ml) and selective during cytotoxicity evaluation.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
SAFC
HEPES
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%