  • Search Results
  • Captopril-polyethyleneimine conjugate modified gold nanoparticles for co-delivery of drug and gene in anti-angiogenesis breast cancer therapy.
Captopril-polyethyleneimine conjugate modified gold nanoparticles for co-delivery of drug and gene in anti-angiogenesis breast cancer therapy.
Journal of biomaterials science. Polymer edition (2015-07-15)
Manhong Li, Yong Li, Xiaohui Huang, Xizhi Lu
ABSTRACT
Captopril-polyethyleneimine (CP) containing low molecular weight polyethyleneimine and anti-angiogenesis drug captopril conjugated via an amide bond was fabricated to modify gold nanoparticles and complex with siRNA to construct siRNA/CP/GNP complexes for the co-delivery of drug and siRNA in anti-angiogenesis breast cancer therapy. The self-assembled siRNA/CP/GNP complexes exhibited desirable and homogenous particle size, reasonable positive charges and condensation ability, and effective gene-silencing property in vitro. In addition, siRNA/CP/GNP complexes co-delivering captopril and siRNA achieved combined angiogenesis suppression by more effectively downregulating the expression of vascular endothelial growth factor mRNA and protein via different pathways in vitro, as compared to mono-delivery systems. In vivo investigation on nude mice bearing MDA-MB435 tumor xenografts revealed that siRNA/CP/GNP complexes possessed satisfying tumor homing ability and strong antitumor activity. These findings suggested that siRNA/CP/GNP complexes could be an ideal system for simultaneous transfer of drug and siRNA, which might be a new promising strategy for effective breast cancer therapy.
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human VEGFA
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Captopril, meets USP testing specifications
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D