MilliporeSigma
  • Doubly end-on azido bridged mixed-valence cobalt trinuclear complex: Spectral study, VTM, inhibitory effect and antimycobacterial activity on human carcinoma and tuberculosis cells.
Doubly end-on azido bridged mixed-valence cobalt trinuclear complex: Spectral study, VTM, inhibitory effect and antimycobacterial activity on human carcinoma and tuberculosis cells.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2015-05-02)
Amitabha Datta, Kuheli Das, Chandana Sen, Nirmal Kumar Karan, Jui-Hsien Huang, Chia-Her Lin, Eugenio Garribba, Chittaranjan Sinha, Tulin Askun, Pinar Celikboyun, Sandeep B Mane
ABSTRACT
Doubly end-on azido-bridged mixed-valence trinuclear cobalt complex, [Co3(L)2(N3)6(CH3OH)2] (1) is afforded by employing a potential monoanionic tetradentate-N2O2 Schiff base precursor (2-[{[2-(dimethylamino)ethyl]imino}methyl]-6-methoxyphenol; HL). Single crystal X-ray structure reveals that in 1, the adjacent Co(II) and Co(III) ions are linked by double end-on azido bridges and thus the full molecule is generated by the site symmetry of a crystallographic twofold rotation axis. Complex 1 is subjected on different spectral analysis such as IR, UV-vis, emission and EPR spectroscopy. On variable temperature magnetic study, we observe that during cooling, the χMT values decrease smoothly until 15K and then reaches to the value 1.56 cm(3) K mol(-1) at 2 K. Complex 1 inhibits the cell growth on human lung carcinoma (A549 cells), human colorectal (COLO 205 and HT-29 cells), and human heptacellular (PLC5 cells) carcinoma cells. Complex 1 exhibits anti-mycobacterial activity and considerable efficacy on Mycobacterium tuberculosis H37Rv ATCC 27294 and H37Ra ATCC 25177 strains.
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sodium azide, SAJ first grade, ≥97.0%
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra