  • Search Results
  • Preclinical Evaluation of DMA, a Bisbenzimidazole, as Radioprotector: Toxicity, Pharmacokinetics, and Biodistribution Studies in Balb/c Mice.
Preclinical Evaluation of DMA, a Bisbenzimidazole, as Radioprotector: Toxicity, Pharmacokinetics, and Biodistribution Studies in Balb/c Mice.
Molecular pharmacology (2015-08-05)
Hemlata Nimesh, Vinod Tiwari, Chunhua Yang, Sushma R Gundala, Krishna Chuttani, Puja P Hazari, Anil K Mishra, Abhisheak Sharma, Jawahar Lal, Anju Katyal, Ritu Aneja, Vibha Tandon
ABSTRACT
Radiotherapy, a therapeutic modality of cancer treatment, nonselectively damages normal tissues as well as tumor tissues. The search is ongoing for therapeutic agents that selectively reduce radiation-induced normal tissue injury without reducing tumoricidal effect, thereby increasing the therapeutic ratio of radiation therapy. Our laboratory established 5-(4-methylpiperazin-1-yl)-2-[2'-(3,4-dimethoxyphenyl)-5'benzimidazolyl] benzimidazole (DMA) as noncytotoxic radioprotector in mammalian cells. DMA showed an excellent radioprotection in mice at single nontoxic oral dose by a dose-reduction factor of 1.28. An oxygen radical absorbing capacity assay confirmed its free-radical quenching ability. Single bolus dose and 28-days of repeated administration of DMA in mice for toxicity studies determined an LD50 of >2000 mg/kg body weight (bw) and 225 mg/kg bw, respectively, suggesting DMA is safe. Histopathology, biochemical parameters, and relative organ weight analysis revealed insignificant changes in the DMA-treated animals. The pharmacokinetic study of DMA at oral and intravenous doses showed its C(max) = 1 hour, bioavailability of 8.84%, elimination half-life of 4 hours, and an enterohepatic recirculation. Biodistribution study in mice with Ehrlich ascites tumors showed that (99m)Tc-DMA achieved its highest concentration in 1 hour and was retained up to 4 hours in the lungs, liver, kidneys, and spleen, and in a low concentration in the tumor, a solicited property of any radioprotector to protect normal cells over cancerous cells. We observed that the single-dose treatment of tumor-bearing mice with DMA 2 hours before 8 Gy total body irradiation showed an impressive rescue of radiation-induced morbidity in terms of weight loss and mortality without a change in tumor response.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
SAFC
Cholesterol, Plant-Derived
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.995% trace metals basis
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.99% trace metals basis
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC