  • Search Results
  • Luminescent determination of quinolones in milk samples by liquid chromatography/post-column derivatization with terbium oxide nanoparticles.
Luminescent determination of quinolones in milk samples by liquid chromatography/post-column derivatization with terbium oxide nanoparticles.
Journal of chromatography. A (2015-06-17)
G S Yánez-Jácome, M P Aguilar-Caballos, A Gómez-Hens
ABSTRACT
The usefulness of terbium oxide nanoparticles (Tb4O7NPs) as post-column derivatizing reagent for the liquid chromatographic determination of residues of quinolone antibiotics in milk samples has been studied. Seven quinolones of veterinary use have been chosen as model analytes to develop this method. The derivatization step is based on the formation of luminescent chelates of quinolones with Tb4O7NPs, which are monitored at λex=340nm and λem=545nm. Another relevant feature of the method is that the use of a 10-cm column and a ternary mixture of methanol, acetonitrile and acetic acid as mobile phase in gradient elution mode allow the chromatographic separation of the quinolones in about 13min, whereas previously described chromatographic methods require about 20min. The dynamic ranges of the calibration graphs and limits of detection are, respectively: 65-900ngmL(-1) and 35ngmL(-1) for marbofloxacin, 7.2-900ngmL(-1) and 2.5ngmL(-1) for ciprofloxacin, 6-900ngmL(-1) and 2ngmL(-1) for danofloxacin, 50-900ngmL(-1) and 20ngmL(-1) for enrofloxacin, 35-900ngmL(-1) and 12ngmL(-1) for sarafloxacin, 5-900ngmL(-1) and 2ngmL(-1) for oxolinic acid, and 7-900ngmL(-1) and 2.5ngmL(-1) for flumequine. The precision, established at two concentration levels of each analyte and expressed as the percentage of the relative standard deviation is in the range of 1.9-8.1% using standards, and of 3.4-10.7% in the presence of milk samples. The method has been satisfactorily applied to the analysis of skimmed, semi-skimmed and whole milk samples, with recoveries ranging from 89.0 to 106.5%.
Sigma-Aldrich
Oxolinic acid, quinolone antibiotic
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Hexamethylenetetramine, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Trioctylphosphine oxide, technical grade, 90%
Sigma-Aldrich
Hexamethylenetetramine, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Trioctylphosphine oxide, ReagentPlus®, 99%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Hexamethylenetetramine, meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hexamethylenetetramine, puriss. p.a., reag. Ph. Eur., ≥99.5% (calc. to the dried substance)
Sigma-Aldrich
Ethanolamine, ≥98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC