  • Search Results
  • Efficient Delivery of DOX to Nuclei of Hepatic Carcinoma Cells in the Subcutaneous Tumor Model Using pH-Sensitive Pullulan-DOX Conjugates.
Efficient Delivery of DOX to Nuclei of Hepatic Carcinoma Cells in the Subcutaneous Tumor Model Using pH-Sensitive Pullulan-DOX Conjugates.
ACS applied materials & interfaces (2015-07-04)
Huanan Li, Yani Cui, Junhui Sui, Shaoquan Bian, Yong Sun, Jie Liang, Yujiang Fan, Xingdong Zhang
ABSTRACT
A series of pullulan-doxorubicin conjugates (Pu-DOXs) were investigated for effectively delivering DOX to nuclei of hepatic carcinoma cells in subcutaneous tumor model. These Pu-DOXs were prepared by conjugating DOX onto pullulan molecule via pH-responsive hydrazone bond using spacers with different alkane chain length. The highest drug loading content of Pu-DOXs went up to nearly 50%, and the diameter of Pu-DOX nanoparticles ranged from 50 to 170 nm, as measured by DLS and TEM. These Pu-DOX nanoparticles could rapidly release DOX in the acidic environment at pH = 5.0 while being kept relatively stable in neural conditions. The in vitro cell coculture experiments revealed that these Pu-DOX nanoparticles were selectively internalized by hepatic carcinoma cells through receptor-mediated endocytosis via asialoglycoprotein receptor on the hepatic carcinoma cell surface. DOX was rapidly released from Pu-DOX nanoparticles in acidic endosome/lysosome, diffused into cell nuclei due to its strong affinity to nucleic acid, inhibited the cell proliferation, and accelerated the cell apoptosis. In the nude mice subcutaneous hepatic carcinoma model, Pu-DOX nanoparticles efficiently accumulated in the tumor site through the enhanced permeation and retention effect. Then DOX was specifically internalized by hepatic carcinoma cells and rapidly diffused into the nuclei of cells. Compared with the control group in in vivo experiments, these Pu-DOX nanoparticles effectively inhibited solid tumor growth, prolonging the lifetime of the experimental animal. These pH sensitive nanoparticles might provide an important clinical implication for targeted hepatic carcinoma therapy with high efficiency and low systematic toxicity.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, 98%
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Hydrazine hydrate, reagent grade, N2H4 50-60 %
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Cytidine 5′-monophosphate, Sigma Grade, ≥99% (HPLC), synthetic, powder
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Oxalyldihydrazide, 98%
Sigma-Aldrich
Succinic dihydrazide, 96%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard