  • Search Results
  • Spectrum-Dependent Spiro-OMeTAD Oxidization Mechanism in Perovskite Solar Cells.
Spectrum-Dependent Spiro-OMeTAD Oxidization Mechanism in Perovskite Solar Cells.
ACS applied materials & interfaces (2015-10-22)
Shen Wang, Wen Yuan, Ying Shirley Meng
ABSTRACT
We propose a spectrum-dependent mechanism for the oxidation of 2,2',7,7'-tetrakis(N,N-di-p-methoxyphenylamine)-9,9'-spirobifluorene (Spiro-OMeTAD) with bis(trifluoromethane)sulfonimide lithium salt (LiTFSI), which is commonly used in perovskite solar cells as the hole transport layer. The perovskite layer plays different roles in the Spiro-OMeTAD oxidization for various spectral ranges. The effect of oxidized Spiro-OMeTAD on the solar cell performance was observed and characterized. With the initial long-wavelength illumination (>450 nm), the charge recombination at the TiO2/Spiro-OMeTAD interface was increased due to the higher amount of the oxidized Spiro-OMeTAD. On the other hand, the increased conductivity of the Spiro-OMeTAD layer and enhanced charge transfer at the Au/Spiro-OMeTAD interface facilitated the solar cell performance.
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%