  • Search Results
  • Changes in signal molecules and maturation promoting factor levels associate with spontaneous resumption of meiosis in rat oocytes.
Changes in signal molecules and maturation promoting factor levels associate with spontaneous resumption of meiosis in rat oocytes.
Cell biology international (2015-01-22)
Shilpa Prasad, Meenakshi Tiwari, Anima Tripathi, Ashutosh N Pandey, Shail K Chaube
ABSTRACT
The present study was aimed to find out changes in signal molecules and maturation promoting factor (MPF) levels during meiotic cell cycle progression from diplotene and metaphase-II (M-II) arrest, a period during which oocyte achieves meiotic competency. Data suggest that high levels of adenosine 3'-5'-cyclic monophosphate (cAMP), guanosine 3'-5'-cyclic monophosphate (cGMP), and nitric oxide (NO) are associated with diplotene arrest, while reduction in their levels correlates with reduced MPF level and meiotic resumption from diplotene arrest. On the other hand, increased intracellular NO, calcium (Ca(2+) ) as well as hydrogen peroxide (H2 O2 ) levels correlate with decreased cAMP, Thr-161 phosphorylated cyclin-dependent kinase-1 (Cdk1) as well as cyclin B1 levels. The decreased Thr-161 phosphorylated Cdk1 and cyclin B1 level reduce MPF level leading to exit from M-II arrest in oocytes cultured in vitro. These data suggest that the decrease of cAMP level and increase of cytosolic free Ca(2+) as well as H2 O2 levels associate with the reduced MPF level and meiotic resumption from diplotene arrest. On the other hand, increase of NO, cGMP, Ca(2+) as well as H2 O2 levels are associated with reduced MPF and spontaneous exit from M-II arrest in rat oocytes cultured in vitro.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate sodium salt, ≥99% (HPLC), powder
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder