  • Search Results
  • MIST, a Novel Approach to Reveal Hidden Substrate Specificity in Aminoacyl-tRNA Synthetases.
MIST, a Novel Approach to Reveal Hidden Substrate Specificity in Aminoacyl-tRNA Synthetases.
PloS one (2015-06-13)
Gilbert Eriani, Joseph Karam, Jomel Jacinto, Erin Morris Richard, Renaud Geslain
ABSTRACT
Aminoacyl-tRNA synthetases (AARSs) constitute a family of RNA-binding proteins, that participate in the translation of the genetic code, by covalently linking amino acids to appropriate tRNAs. Due to their fundamental importance for cell life, AARSs are likely to be one of the most ancient families of enzymes and have therefore been characterized extensively. Paradoxically, little is known about their capacity to discriminate tRNAs mainly because of the practical challenges that represent precise and systematic tRNA identification. This work describes a new technical and conceptual approach named MIST (Microarray Identification of Shifted tRNAs) designed to study the formation of tRNA/AARS complexes independently from the aminoacylation reaction. MIST combines electrophoretic mobility shift assays with microarray analyses. Although MIST is a non-cellular assay, it fully integrates the notion of tRNA competition. In this study we focus on yeast cytoplasmic Arginyl-tRNA synthetase (yArgRS) and investigate in depth its ability to discriminate cellular tRNAs. We report that yArgRS in submicromolar concentrations binds cognate and non-cognate tRNAs with a wide range of apparent affinities. In particular, we demonstrate that yArgRS binds preferentially to type II tRNAs but does not support their misaminoacylation. Our results reveal important new trends in tRNA/AARS complex formation and potential deep physiological implications.
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Spermidine, BioReagent, for molecular biology, suitable for cell culture, ≥98%
Sigma-Aldrich
Spermidine, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 99.99 atom % 14N, 40 wt. % in H2O