  • Search Results
  • Clinicopathological characteristics and outcomes of ROS1-rearranged patients with lung adenocarcinoma without EGFR, KRAS mutations and ALK rearrangements.
Clinicopathological characteristics and outcomes of ROS1-rearranged patients with lung adenocarcinoma without EGFR, KRAS mutations and ALK rearrangements.
Thoracic cancer (2015-08-15)
Shafei Wu, Jinghui Wang, Lijuan Zhou, Dan Su, Yuanyuan Liu, Xiaolong Liang, Shucai Zhang, Xuan Zeng
ABSTRACT
c-ros oncogene 1 (ROS1) rearrangement presents one of the newest molecular targets in non-small cell lung cancer (NSCLC). ROS1 rearrangement is predominantly found in adenocarcinoma cases and is exclusive to other oncogenes, such as epidermal growth factor receptor (EGFR), Kirsten rat sarcoma viral oncogene homolog (KRAS), and anaplastic lymphoma kinase (ALK). The aim of this study was to investigate the clinicopathological characteristics and outcomes of ROS1-rearranged patients with lung adenocarcinoma without EGFR and KRAS mutations and ALK rearrangements. Wild-type EGFR/KRAS/ALK patients with lung adenocarcinoma were selected from Beijing Chest Hospital. Specimens were conducted in tissue microarrays. ROS1 rearrangement was screened using fluorescence in situ hybridization. Our study included 127 patients with lung adenocarcinoma without EGFR and KRAS mutations and ALK rearrangements. ROS1 rearrangement was detected in five (3.9%) of the 127 patients. Compared with ROS1-negative patients, the positive rate of ROS1 in female patients was significantly higher than in male patients (9.8% vs. 0.0%, P = 0.009). There were no differences in age, smoking status, stage or histological subtype between ROS1-positive and ROS1-negative patients. No significant difference in survival was detected between the ROS1-positive and ROS1-negative patients. ROS1 rearrangement is a rare subset of lung adenocarcinoma. In 127 patients with lung adenocarcinoma, 3.9% of ROS1-positive patients with wild-type EGFR/KRAS/ALK were found.
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane