  • Search Results
  • Reversible deformation-formation of a multistimuli responsive vesicle by a supramolecular peptide amphiphile.
Reversible deformation-formation of a multistimuli responsive vesicle by a supramolecular peptide amphiphile.
Soft matter (2015-05-27)
Julfikar Hassan Mondal, Sahnawaz Ahmed, Titli Ghosh, Debapratim Das
ABSTRACT
A systematic study of the ternary complex formation process for aromatic amino acids using ucurbit[8]uril (CB[8]) and a viologen amphiphile shows that the affinity of the amino acid needs to be higher or in a comparable range to that of CB[8] for the amphiphile in order to form the ternary complex. Based on these observations, a supramolecular peptide amphiphile and its corresponding vesicle are prepared using a peptide containing an azobenzene moiety. The azobenzene group at the N-terminus of the peptide served as the second guest for CB[8]. The vesicles obtained from this peptide amphiphile show response to a number of external triggers. The trans-cis isomerization of the azobenzene group upon irradiation with UV-light of 365 nm leads to the breakdown of the ternary complex and eventually to the disruption of the vesicle. The deformation-reformation of the vesicle can be controlled by illuminating the disrupted solution with light of 420 nm as it facilitates the cis-trans isomerization. Thus, the vesicle showed a controlled and reversible response to UV-light with the ability for manipulation of the formation-deformation of the vesicle by the choice of an appropriate wavelength. The vesicle showed response to a stronger guest (1-adamantylamine) for CB[8], which displaces both the guests from the CB[8] cavity and consequently ruptures the vesicle structure. 2,6-Dihydroxynaphthalene acts as a competitive guest and thereby behaves as another external trigger for replacing the peptide from the CB[8] cavity by self-inclusion to form the ternary complex. Henceforth, it allows retaining the vesicle structure and results in the release of the peptide from the vesicle.
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
5(6)-Carboxyfluorescein, Dye content 90 %
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
1-Bromohexadecane, 97%
Sigma-Aldrich
5(6)-Carboxyfluorescein, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D