  • Search Results
  • Application of passive sampling for measuring dissolved concentrations of organic contaminants in the water column at three marine superfund sites.
Application of passive sampling for measuring dissolved concentrations of organic contaminants in the water column at three marine superfund sites.
Environmental toxicology and chemistry (2015-06-04)
Robert M Burgess, Rainer Lohmann, Joseph P Schubauer-Berigan, Pamela Reitsma, Monique M Perron, Lisa Lefkovitz, Mark G Cantwell
ABSTRACT
Currently, there is an effort under way to encourage remedial project managers at contaminated sites to use passive sampling to collect freely dissolved concentrations (Cfree ) of hydrophobic organic contaminants to improve site assessments. The objective of the present study was to evaluate the use of passive sampling for measuring water column Cfree for several hydrophobic organic contaminants at 3 US Environmental Protection Agency Superfund sites. Sites investigated included New Bedford Harbor (New Bedford, MA, USA), Palos Verdes Shelf (Los Angeles, CA, USA), and Naval Station Newport (Newport, RI, USA); and the passive samplers evaluated were polyethylene, polydimethylsiloxane-coated solid-phase microextraction fibers, semipermeable membrane devices, and polyoxymethylene. In general, the different passive samplers demonstrated good agreement, with Cfree values varying by a factor of 2 to 3. Further, at New Bedford Harbor, where conventional water sample concentrations were also measured (i.e., grab samples), passive sampler-based Cfree values agreed within a factor of 2. These findings suggest that all of the samplers were experiencing and measuring similar Cfree during their respective deployments. Also, at New Bedford Harbor, a strong log-linear, correlative, and predictive relationship was found between polyethylene passive sampler accumulation and lipid-normalized blue mussel bioaccumulation of polychlorinated biphenyls (r(2)  = 0.92, p < 0.05). The present study demonstrates the utility of passive sampling for generating scientifically accurate water column Cfree values, which is critical for making informed environmental management decisions at contaminated sediment sites.
Sigma-Aldrich
Glyceryl trioleate, ~65%
Sigma-Aldrich
Glyceryl trioleate, ≥99%
Sigma-Aldrich
Glyceryl trioleate, ≥97.0% (TLC)
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with autosampler, needle size 24 ga
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with autosampler, needle size 24 ga
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm (PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with manual holder, needle size 24 ga
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm (PDMS), needle size 23 ga, for use with autosampler
Supelco
SPME-OC Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 75 μm (Coating thickness includes 65 μm coating + 10 μm OC (overcoating)), needle size 23 ga, pkg of 3 ea, fiber L 1 cm
Supelco
IonSense® C18 SPE-it Tips, pkg of 96 ea
Supelco
SPME LC Tips, PDMS/DVB, pkg of 96 ea
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Supelco
IonSense® PDMS/DVB SPE-it Tips, pkg of 96 ea
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (PDMS/DVB), for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm (PEG), metal alloy fiber, for use with manual holder
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm (PEG), for use with autosampler
Supelco
SPME LC Tips, C18, tray of 96 ea