  • Search Results
  • Computational redesign of the lipid-facing surface of the outer membrane protein OmpA.
Computational redesign of the lipid-facing surface of the outer membrane protein OmpA.
Proceedings of the National Academy of Sciences of the United States of America (2015-07-23)
James A Stapleton, Timothy A Whitehead, Vikas Nanda
ABSTRACT
Advances in computational design methods have made possible extensive engineering of soluble proteins, but designed β-barrel membrane proteins await improvements in our understanding of the sequence determinants of folding and stability. A subset of the amino acid residues of membrane proteins interact with the cell membrane, and the design rules that govern this lipid-facing surface are poorly understood. We applied a residue-level depth potential for β-barrel membrane proteins to the complete redesign of the lipid-facing surface of Escherichia coli OmpA. Initial designs failed to fold correctly, but reversion of a small number of mutations indicated by backcross experiments yielded designs with substitutions to up to 60% of the surface that did support folding and membrane insertion.
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
DL-Phenylalanine, ReagentPlus®, 99%
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Anti-HA−FITC antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture