  • Search Results
  • Curcumin retunes cholesterol transport homeostasis and inflammation response in M1 macrophage to prevent atherosclerosis.
Curcumin retunes cholesterol transport homeostasis and inflammation response in M1 macrophage to prevent atherosclerosis.
Biochemical and biophysical research communications (2015-10-17)
Fang-Yuan Chen, Juan Zhou, Ning Guo, Wang-Ge Ma, Xin Huang, Huan Wang, Zu-Yi Yuan
ABSTRACT
Lipoprotein cholesterol metabolism dysfunction in the arterial wall is a major contributor to atherosclerosis, and excessive lipid intake and failed cholesterol homeostasis may accelerate the atherogenic process. Curcumin exerts multiple effects by alleviating inflammation, hyperlipidemia, and atherosclerosis; however, its role in cholesterol transport homeostasis and its underlying impact on inflammatory M1 macrophages are poorly understood. This work aimed to investigate the effect of curcumin on cholesterol transport, the inflammatory response and cell apoptosis in M1 macrophages. RAW264.7 macrophages (M0) were induced with LPS plus IFN-γ for 12 h to develop a M1 subtype and were then incubated with curcumin at different concentrations (6.25 and 12.5 μmol/L) in the presence or absence of oxLDL. Then, cholesterol influx/efflux and foam cell formation as well as inflammation and apoptosis were evaluated. It was found that curcumin increased cholesterol uptake measured by the Dil-oxLDL binding assay, and simultaneously increased cholesterol efflux carried out by Apo-A1 and HDL in M1 cells. Curcumin further reinforced ox-LDL-induced cholesterol esterification and foam cell formation as determined by Oil Red O and BODIPY staining. Moreover, curcumin dramatically reduced ox-LDL-induced cytokine production such as IL-1β, IL-6 as well as TNF-α and M1 cell apoptosis. We also found that curcumin upregulated CD36 and ABCA1 in M1 macrophages. Curcumin increased PPARγ expression, which in turn promoted CD36 and ABCA1 expression. In conclusion, curcumin may increase the ability of M1 macrophages to handle harmful lipids, thus promoting lipid processing, disposal and removal, which may support cholesterol homeostasis and exert an anti-atherosclerotic effect.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
SAFC
Cholesterol, Plant-Derived
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
GW9662, >98% (HPLC)
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
2-Chloro-5-nitrobenzanilide, 97%