  • Search Results
  • Elicitation of T-cell responses by structural and non-structural proteins of coxsackievirus B4.
Elicitation of T-cell responses by structural and non-structural proteins of coxsackievirus B4.
The Journal of general virology (2014-11-09)
Suvi Bengs, Jane Marttila, Petri Susi, Jorma Ilonen
ABSTRACT
Coxsackievirus B4 (CV-B4) belongs to the genus Enterovirus within the family Picornaviridae. To investigate target proteins recognized by T-cells in human enterovirus B infections, virus-encoded structural [VP0 (VP4 and VP2), VP1, VP3] and non-structural (2A, 2B, 2C, 3C and 3D) proteins were expressed and purified in Escherichia coli. Peripheral blood of 19 healthy adult donors was used to create enterovirus-specific T-cell lines by repeated stimulation with CV-B4 cell lysate antigen. T-cell lines responded in individual patterns, and responses to all purified proteins were observed. The most often recognized enteroviral protein was VP0, which is the fusion between the most conserved structural proteins, VP4 and VP2. T-cell responses to VP0 were detected in 15 of the 19 (79 %) donor lines. Non-structural 2C protein was recognized in 11 of the 19 (58 %) lines, and 11 of the 19 (58 %) lines also had a response to 3D protein. Furthermore, responses to other non-structural proteins (2A, 2B and 3C) were also detected. T-cell responses did not correlate clearly to the individual HLA-DR-DQ phenotype or the history of past coxsackie B virus infections of the donors.
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems
Sigma-Aldrich
IL-2 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)