  • Search Results
  • s-Ethyl Cysteine and s-Methyl Cysteine Protect Human Bronchial Epithelial Cells Against Hydrogen Peroxide Induced Injury.
s-Ethyl Cysteine and s-Methyl Cysteine Protect Human Bronchial Epithelial Cells Against Hydrogen Peroxide Induced Injury.
Journal of food science (2015-09-16)
Te-chun Hsia, Mei-chin Yin
ABSTRACT
Protective effects and actions from s-ethyl cysteine (SEC) and s-methyl cysteine (SMC) for BEAS-2B cells were examined. BEAS-2B cells were pretreated with SEC or SMC at 4, 8, or 16 μmol/L, and followed by hydrogen peroxide (H2 O2 ) treatment. Data showed that H2 O2 enhanced Bax, caspase-3 and caspase-8 expression, and declined Bcl-2 expression. However, SEC or SMC dose-dependently decreased caspase-3 expression and reserved Bcl-2 expression. H2 O2 increased reactive oxygen species (ROS) production, and lowered glutathione level, glutathione peroxide, and glutathione reductase activities in BEAS-2B cells. SEC or SMC pretreatments reduced ROS generation, and maintained glutathione redox cycle in those cells. H2 O2 upregulated the expression of both p47(phox) and gp91(phox) . SEC and SMC downregulated p47(phox) expression. SEC or SMC at 8 and 16 μmol/L decreased H2 O2 -induced release of inflammatory cytokines. H2 O2 stimulated the activation of nuclear factor-κB (NF-κB) and mitogen-activated protein kinase. SEC and SMC pretreatments dose-dependently downregulated NF-κB p65 and p-p38 expression. Pyrrolidine dithiocarbamate or SB203580 inhibited NF-κB activation and p38 phosphorylation; thus, SEC or SMC pretreatments failed to affect protein expression of these factors. These novel findings suggest that SEC or SMC could protect bronchial cells and benefit respiratory epithelia stability and functions.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
MTT Formazan, powder
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
MTT Formazan, MTT reduction end product