  • Search Results
  • Monitoring tryptophan metabolism after exposure to hexaconazole and the enantioselective metabolism of hexaconazole in rat hepatocytes in vitro.
Monitoring tryptophan metabolism after exposure to hexaconazole and the enantioselective metabolism of hexaconazole in rat hepatocytes in vitro.
Journal of hazardous materials (2015-04-13)
Yao Wang, Wentao Zhu, Jing Qiu, Xinru Wang, Ping Zhang, Jin Yan, Zhiqiang Zhou
ABSTRACT
In the present study, the enantioselective metabolism, cytotoxicity of hexaconazole and its influence on tryptophan metabolism in rat hepatocytes in vitro were investigated. Following the exposure of primary rat hepatocytes to rac-hexaconazole, the concentrations of its enantiomers in the media were determined by chiral high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS). The half-lives (t1/2) of (+)-hexaconazole and (-)-hexaconazole were 5.17 h and 19.80 h, respectively, indicating that the metabolic process was enantioselective with (-)-hexaconazole enrichment. Using the MTT method, the EC50 values of rac-hexaconazole, (+)-hexaconazole and (-)-hexaconazole after 12h of exposure were determined to be 71.62, 62.71 and 67.94 μM, respectively. Tryptophan metabolism was monitored using metabolomics profiling techniques. Hexaconazole and its enantiomers caused the down-regulation of tryptophan levels and the up-regulation of kynurenine (KYN) levels, suggesting a role for hexaconazole in the activation of the KYN pathway and providing information for the mechanism of its toxicity.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
SAFC
HEPES
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
DL-Indole-3-lactic acid, 99%
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
5-Hydroxyindole-3-acetic acid, ≥98% (HPLC), crystalline
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
3-Hydroxy-DL-kynurenine
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
3-Indolepropionic acid, ReagentPlus®, 99%
Sigma-Aldrich
3-Hydroxyanthranilic acid, 97%
Sigma-Aldrich
Indole-3-propionic acid, ≥99.0% (T)