  • Search Results
  • A novel function of nuclear nonmuscle myosin regulatory light chain in promotion of xanthine oxidase transcription after myocardial ischemia/reperfusion.
A novel function of nuclear nonmuscle myosin regulatory light chain in promotion of xanthine oxidase transcription after myocardial ischemia/reperfusion.
Free radical biology & medicine (2015-02-24)
Yi-Shuai Zhang, Bin Liu, Xiu-Ju Luo, Jie-Jie Zhang, Nian-Sheng Li, Qi-Lin Ma, Jun-Lin Jiang, Yuan-Jian Li, Qingjie Li, Jun Peng
ABSTRACT
Nuclear myosin regulates gene transcription and this novel function might be modulated through phosphorylation of the myosin regulatory light chain (p-MLC20). Nonmuscle MLC20 (nmMLC20) is also present in the nuclei of cardiomyocytes and a potential nmMLC20 binding sequence has been identified in the promoter of the xanthine oxidase (XO) gene. Thus, we investigated its function in the regulation of XO transcription after myocardial ischemia/reperfusion (IR). In a rat model of myocardial IR and a cardiomyocyte model of hypoxia/reoxygenation (HR) injury, the cardiac or cell injury, myosin light chain kinase (MLCK) content, XO expression and activity, XO-derived products, and level of nuclear p-nmMLC20 were detected. Coimmunoprecipitation (co-IP), chromatin immunoprecipitation, DNA pull-down, and luciferase reporter gene assays were used to decipher the molecular mechanisms through which nmMLC20 promotes XO expression. IR or HR treatment dramatically elevated nuclear p-nmMLC20 level, accompanied by increased XO expression, activity, and products (H2O2 and uric acid), as well as the IR or HR injury; these effects were ameliorated by inhibition of MLCK or knockdown of nmMLC20. Our findings from these experiments demonstrated that nuclear p-nmMLC20 binds to the consensus sequence GTCGCC in the XO gene promoter, interacts with RNA polymerase II and transcription factor IIB to form a transcription preinitiation complex, and hence activates XO gene transcription. These results suggest that nuclear p-nmMLC20 plays an important role in IR/HR injury by transcriptionally upregulating XO gene expression to increase oxidative stress in myocardium. Our findings demonstrate nuclear nmMLC20 as a potential new therapeutic target to combat cardiac IR injury.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate