  • Search Results
  • Age-dependent uncoupling of mitochondria from Ca2⁺ release units in skeletal muscle.
Age-dependent uncoupling of mitochondria from Ca2⁺ release units in skeletal muscle.
Oncotarget (2015-10-21)
Laura Pietrangelo, Alessandra D'Incecco, Alina Ainbinder, Antonio Michelucci, Helmut Kern, Robert T Dirksen, Simona Boncompagni, Feliciano Protasi
ABSTRACT
Calcium release units (CRUs) and mitochondria control myoplasmic [Ca2+] levels and ATP production in muscle, respectively. We recently reported that these two organelles are structurally connected by tethers, which promote proximity and proper Ca2+ signaling.Here we show that disposition, ultrastructure, and density of CRUs and mitochondria and their reciprocal association are compromised in muscle from aged mice. Specifically, the density of CRUs and mitochondria is decreased in muscle fibers from aged (>24 months) vs. adult (3-12 months), with an increased percentage of mitochondria being damaged and misplaced from their normal triadic position. A significant reduction in tether (13.8 ± 0.4 vs. 5.5 ± 0.3 tethers/100 µm2) and CRU-mitochondrial pair density (37.4 ± 0.8 vs. 27.0 ± 0.7 pairs/100 µm2) was also observed in aged mice. In addition, myoplasmic Ca2+ transient (1.68 ± 0.08 vs 1.37 ± 0.03) and mitochondrial Ca2+ uptake (9.6 ± 0.050 vs 6.58 ± 0.54) during repetitive high frequency tetanic stimulation were significantly decreased. Finally oxidative stress, assessed from levels of 3-nitrotyrosine (3-NT), Cu/Zn superoxide-dismutase (SOD1) and Mn superoxide dismutase (SOD2) expression, were significantly increased in aged mice. The reduced association between CRUs and mitochondria with aging may contribute to impaired cross-talk between the two organelles, possibly resulting in reduced efficiency in activity-dependent ATP production and, thus, to age-dependent decline of skeletal muscle performance.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Sodium chloride, tablet