  • Search Results
  • A new role for an old drug: Ambroxol triggers lysosomal exocytosis via pH-dependent Ca²⁺ release from acidic Ca²⁺ stores.
A new role for an old drug: Ambroxol triggers lysosomal exocytosis via pH-dependent Ca²⁺ release from acidic Ca²⁺ stores.
Cell calcium (2015-11-13)
Giorgio Fois, Nina Hobi, Edward Felder, Andreas Ziegler, Pika Miklavc, Paul Walther, Peter Radermacher, Thomas Haller, Paul Dietl
ABSTRACT
Ambroxol (Ax) is a frequently prescribed drug used to facilitate mucociliary clearance, but its mode of action is yet poorly understood. Here we show by X-ray spectroscopy that Ax accumulates in lamellar bodies (LBs), the surfactant storing, secretory lysosomes of type II pneumocytes. Using lyso- and acidotropic substances in combination with fluorescence imaging we confirm that these vesicles belong to the class of acidic Ca(2+) stores. Ax lead to a significant neutralization of LB pH, followed by intracellular Ca(2+) release, and to a dose-dependent surfactant exocytosis. Ax-induced Ca(2+) release was significantly reduced and slowed down by pretreatment of the cells with bafilomycin A1 (Baf A1), an inhibitor of the vesicular H(+) ATPase. These results could be nearly reproduced with NH3/NH4(+). The findings suggest that Ax accumulates within LBs and severely affects their H(+) and Ca(2+) homeostasis. This is further supported by an Ax-induced change of nanostructural assembly of surfactant layers. We conclude that Ax profoundly affects LBs presumably by disordering lipid bilayers and by acting as a weak base. The pH change triggers - at least in part - Ca(2+) release from stores and secretion of surfactant from type II cells. This novel mechanism of Ax as a lysosomal secretagogue may also play a role for its recently discussed use for lysosomal storage and other degenerative diseases.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Fura 2-AM, ≥95% (HPLC)
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Fura 2-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC