  • Search Results
  • Combined treatment with a pH-sensitive fusogenic peptide and cationic lipids achieves enhanced cytosolic delivery of exosomes.
Combined treatment with a pH-sensitive fusogenic peptide and cationic lipids achieves enhanced cytosolic delivery of exosomes.
Scientific reports (2015-05-27)
Ikuhiko Nakase, Shiroh Futaki
ABSTRACT
Exosomes, which are approximately 100 nm vesicles secreted by cells, have been studied with respect to cell-to-cell communication, disease diagnosis, and intracellular delivery. The cellular uptake of exosomes occurs by endocytosis; however, the cytosolic release efficiency of encapsulated molecules inside cells is low. To address this issue, here we demonstrate a simple technique for enhancing the cellular uptake and cytosolic release of exosomes by combining a pH-sensitive fusogenic peptide for the fusion of endosomal and exosomal membranes inside cells. This method stimulates the efficient cytosolic release of the exosomal contents with cationic lipids that act as a "glue" to support cellular uptake. Using this simple combined technique, the effective cellular uptake and cytosolic release of an artificially encapsulated dextran macromolecule (70 kDa) in exosomes are achieved, and a marked improvement in bioactivity is attained with the artificially encapsulated ribosome-inactivating protein saporin. Our method will contribute to many biological research fields, including the assessment of the activities of exosomal contents and the development of candidate tools enabling intracellular visualisation and cellular regulation for future therapeutic applications.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Water, BioPerformance Certified