  • Search Results
  • Caspase-7: a critical mediator of optic nerve injury-induced retinal ganglion cell death.
Caspase-7: a critical mediator of optic nerve injury-induced retinal ganglion cell death.
Molecular neurodegeneration (2015-08-27)
Shreyasi Choudhury, Yang Liu, Abbot F Clark, Iok-Hou Pang
ABSTRACT
Axonal injury of the optic nerve (ON) is involved in various ocular diseases, such as glaucoma and traumatic optic neuropathy, which leads to apoptotic death of retinal ganglion cells (RGCs) and loss of vision. Caspases have been implicated in RGC pathogenesis. However, the role of caspase-7, a functionally unique caspase, in ON injury and RGC apoptosis has not been reported previously. The purpose of this study is to evaluate the role of caspase-7 in ON injury-induced RGC apoptosis. C57BL/6 (wildtype, WT) and caspase-7 knockout (Casp7(-/-)) mice were used. We show that ON crush activated caspase-7 and calpain-1, an upstream activator of caspase-7, in mouse RGCs, as well as hydrolysis of kinectin and co-chaperone P23, specific substrates of caspase-7. ON crush caused a progressive loss of RGCs to 28 days after injury. Knockout of caspase-7 partially and significantly protected against the ON injury-induced RGC loss; RGC density at 28 days post ON crush in Casp7(-/-) mice was approximately twice of that in WT ON injured retinas. Consistent with changes in RGC counts, spectral-domain optical coherence tomography analysis revealed that ON crush significantly reduced the in vivo thickness of the ganglion cell complex layer (including ganglion cell layer, nerve fiber layer, and inner plexiform layer) in the retina. The ON crush-induced thinning of retinal layer was significantly ameliorated in Casp7(-/-) mice when compared to WT mice. Moreover, electroretinography analysis demonstrated a decline in the positive component of scotopic threshold response amplitude in ON crushed eyes of the WT mice, whereas this RGC functional response was significantly higher in Casp7(-/-) mice at 28 days post injury. Altogether, our findings indicate that caspase-7 plays a critical role in ON injury-induced RGC death, and inhibition of caspase-7 activity may be a novel therapeutic strategy for glaucoma and other neurodegenerative diseases of the retina.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65,000 IU/g
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC