  • Search Results
  • Validation of a High-Performance Liquid Chromatography-Tandem Mass Spectrometry Method for the Determination of Perhexiline and Cis-Hydroxy-Perhexiline Plasma Concentrations.
Validation of a High-Performance Liquid Chromatography-Tandem Mass Spectrometry Method for the Determination of Perhexiline and Cis-Hydroxy-Perhexiline Plasma Concentrations.
Therapeutic drug monitoring (2015-03-17)
Ian S Westley, Giovanni Licari, Benedetta C Sallustio
ABSTRACT
The polymorphic nature of cytochrome P450 2D6 has made therapeutic drug monitoring of the anti-anginal agent perhexiline a compulsory step in reducing adverse events associated with plasma concentrations above the therapeutic range (0.15-0.60 mg/L). The aim of this study was to develop a high-performance liquid chromatography-mass spectrometry/mass spectrometry method for the determination of plasma perhexiline concentrations and its major metabolite cis-hydroxy-perhexiline to reduce sample extraction procedures and improve sample turnaround times. The method was validated by determining the precision and accuracy of calibrators and quality control material, comparing quality assurance program samples and patient samples measured by a previously reported liquid-liquid extraction fluorescence (FL) detection high-performance liquid chromatography method and performing matrix effects investigations. Replicates of calibrators at concentrations of 3.00 and 0.05 mg/L demonstrated imprecision of <10.8% and inaccuracy of <8.2% for perhexiline and <10.1% and <4.5% for cis-hydroxy-perhexiline, respectively. All samples measured by the 2 methods (n = 102) demonstrated Deming regression of perhexiline = 1.20 FL + 0.00 (Sy.x = 0.08, 1/slope = 0.67); cis-hydroxy-perhexiline = 1.48 FL - 0.20 (Sy.x = 0.40, 1/slope = 0.67). The assay performance was deemed acceptable and integrated into the routine therapeutic drug monitoring program of the department.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid