  • Search Results
  • Multifunctional Architectures Constructing of PANI Nanoneedle Arrays on MoS2 Thin Nanosheets for High-Energy Supercapacitors.
Multifunctional Architectures Constructing of PANI Nanoneedle Arrays on MoS2 Thin Nanosheets for High-Energy Supercapacitors.
Small (Weinheim an der Bergstrasse, Germany) (2015-06-06)
Jixin Zhu, Wenping Sun, Dan Yang, Yu Zhang, Hng Huey Hoon, Hua Zhang, Qingyu Yan
ABSTRACT
Multifunctional MoS2 @PANI (polyaniline) pseudo-supercapacitor electrodes consisting of MoS2 thin nanosheets and PANI nanoarrays are fabricated via a large-scale approach. The superior capacitance retention is retained up to 91% after 4000 cycles and a high energy density of 106 Wh kg(-1) is delivered at a power density of 106 kW kg(-1) .
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)