  • Search Results
  • A rapid high-performance liquid chromatography-tandem mass spectrometry assay for unambiguous detection of different milk species employed in cheese manufacturing.
A rapid high-performance liquid chromatography-tandem mass spectrometry assay for unambiguous detection of different milk species employed in cheese manufacturing.
Journal of dairy science (2015-10-12)
Nadia Bernardi, Giuseppe Benetti, Naceur M Haouet, Manuel Sergi, Lisa Grotta, Sonia Marchetti, Federica Castellani, Giuseppe Martino
ABSTRACT
The aim of the study was to investigate the possibility to differentiate the 4 most important species in Italian dairy industry (cow, buffalo, sheep, and goat), applying a bottom-up proteomic approach to assess the milk species involved in cheese production. Selective peptides were detected in milk to use as markers in cheese products. Trypsin-digested milk samples of cow, sheep, goat, and buffalo, analyzed by HPLC-tandem mass spectrometry provided species-specific peptides, some of them recognized by Mascot software (Matrix Science Ltd., Boston, MA) as derived from well-known species specific proteins. A multianalyte multiple reaction monitoring method, built with these specific peptides, was successfully applied to cheeses with different composition, showing high specificity in detection of species involved. Neither aging nor production method seemed to affect the response, demonstrating that chosen peptides well act as species markers for dairy products.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
SAFC
Iodoacetamide
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)