  • Search Results
  • FV-429 Induced Apoptosis Through ROS-Mediated ERK2 Nuclear Translocation and p53 Activation in Gastric Cancer Cells.
FV-429 Induced Apoptosis Through ROS-Mediated ERK2 Nuclear Translocation and p53 Activation in Gastric Cancer Cells.
Journal of cellular biochemistry (2015-02-05)
Yuxin Zhou, Libin Wei, Haiwei Zhang, Qinsheng Dai, Zhiyu Li, Boyang Yu, Qinglong Guo, Na Lu
ABSTRACT
Following our previous finding which revealed that FV-429 induces apoptosis in human hepatocellular carcinoma HepG2 cells, in this study, we found that FV-429 could also induce apoptosis in human gastric cancer cells. Firstly, FV-429 inhibited the viability of BGC-823 and MGC-803 cells with IC50 values in the range of 38.10 ± 6.28 and 31.53 ± 6.84 µM for 24 h treatment by MTT-assay. Secondly, FV-429 induced apoptosis in BGC-823 and MGC-803 cells through the mitochondrial-mediated pathway, showing an increase in Bax/Bcl-2 ratios, and caspase-9 activation, without change in caspase-8. Further research revealed that the mitogen-activated protein kinases, including c-Jun N-terminal kinase, extracellular regulated kinase, and p38 mitogen-activated protein kinase, could be activated by FV-429-induced high level ROS. Moreover, FV-429 also promoted the ERK2 nuclear translocation, resulting in the co-translocation of p53 to the nucleus and increased transcription of p53-regulated proapoptotic genes. FV-429 significantly inhibited the nude mice xenograft tumors growth of BGC-823 or MGC-803 cells in vivo.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)