  • Search Results
  • The fluorescence and infrared absorption probe para-cyanophenylalanine: Effect of labeling on the behavior of different membrane-interacting peptides.
The fluorescence and infrared absorption probe para-cyanophenylalanine: Effect of labeling on the behavior of different membrane-interacting peptides.
Biopolymers (2015-05-15)
Sara Bobone, Marta De Zotti, Annalisa Bortolotti, Barbara Biondi, Gema Ballano, Antonio Palleschi, Claudio Toniolo, Fernando Formaggio, Lorenzo Stella
ABSTRACT
Total syntheses and complete characterizations of singly substituted PheCN -based analogs of alamethicin AlaP, which is active on model and natural membranes, and the TM peptide, which inserts in a transmembrane orientation in lipid bilayers, are reported. The syntheses of the AlaP analogs were performed in solution, while those of TM and its analogs were carried out by solid phase. Using the cyanophenyl fluorescence and infrared (IR) absorption probe, an in-depth investigation of the self-association, membrane-interacting, permeabilizing, and orientation properties of these peptides were conducted. The aromatic residue incorporated induces only a negligible modification to the properties of the parent peptides. The PheCN IR absorption band was located between 2228 and 2230 cm(-1) for all peptides, irrespective of the position of labeling. By contrast, as the width of this band varied significantly with the depth of probe insertion in the bilayer, it could represent a good marker of the PheCN position in phospholipid membranes.
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%
Sigma-Aldrich
HATU, 97%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)