  • Search Results
  • Optimized Treatment of Heparinized Blood Fractions to Make Them Suitable for Analysis.
Optimized Treatment of Heparinized Blood Fractions to Make Them Suitable for Analysis.
Biopreservation and biobanking (2015-08-19)
María Teresa Sánchez-Fito, Elisa Oltra
ABSTRACT
It has been known for decades that many cytokines, such as IL-2, IL-6, and IL-12, bind to heparin. Even though some enzyme-linked immunosorbent assays (ELISA) use antibody-recognizing epitopes not affected by this binding, ELISA manufacturers often warn that heparinized plasma or serum fractions containing more than 3 IU (international units)/mL of heparin should not be used in assays so as to prevent heparin interference in the reaction. In addition, enzyme-based nucleic acid amplifications from heparinized samples have been shown defective by several research groups. The aim of this study was to determine optimal degradation and/or removal of heparin from heparinized blood samples to best turn them into fractions for appropriate ELISA and RT-PCR analysis. A colorimetric reporter assay based on the metachromatic effect of the binding of heparin to toluidine blue was shown to be a low-cost effective method to discriminate assay compatible blood fractions with heparin levels below 3 IU/mL. Heparin removal from human blood fractions was best achieved by treatment with either Bacteroides Heparinase II or the less expensive Heparinase I at a final concentration of 0.1 U/μL and incubations at 30°C for a period between 30 min and 4 h, or by adsorption to Ecteola slurries at a concentration of 20 mg/mL for 20 min at room temperature (RT). The fact that both enzymatic and resin-based optimized treatments allowed for replication of the readings obtained with heparin-free equivalent fractions in both ELISA and RT-PCR assays indicates they should be appropriate for quantitative studies such as expression profiling at both the protein and nucleic acid level. The cost-effective protocols developed in this study could make heparinized, otherwise unusable, blood-derived collections suitable for analysis by ELISA and RT-PCR amplifications, among other analyses, enhancing the possibilities for studying valuable bio-banked heparinized human samples.
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
TRI Reagent®, BD, For processing whole blood, plasma, or serum.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Lithium-7Li, ≥99.8 atom % 7Li, ≥99.8% (CP)
SAFC
Triethanolamine
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Ecteola Cellulose, coarse
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis