  • Search Results
  • Combination of retrograde superselective intra-arterial chemotherapy and Seldinger method in locally advanced oral cancer.
Combination of retrograde superselective intra-arterial chemotherapy and Seldinger method in locally advanced oral cancer.
Journal of cancer research and therapeutics (2015-07-08)
Masataka Uehara, Ryouichi Ohya, Masaaki Kodama, Takeshi Shiraishi, Izumi Asahina, Kazuhiro Tominaga
ABSTRACT
The nonsurgical strategies for locally advanced oral cancer are desirable. Superselective intra-arterial infusion with radiotherapy was utilized for this purpose, and there are two types of superselective intra-arterial infusion methods: The Seldinger method and the retrograde superselective intra-arterial chemotherapy (HFT method). In one case, the HFT method was applied to locally advanced tongue cancer, and the Seldinger method was used for additional administration of cisplatin (CDDP) to compensate for a lack of drug flow in the HFT method. In another case, the HFT method was applied to locally advanced lower gingival cancer. The Seldinger method was applied to metastatic lymph nodes. In both cases, additional administration of CDDP using the Seldinger method resulted in a complete response. The combination of the HFT and Seldinger methods was useful to eradicate locally advanced oral cancer because each method compensated for the defects of the other.
Sigma-Aldrich
Indigo carmine, certified by the Biological Stain Commission, Dye content 85 %
Sigma-Aldrich
Indigo carmine, for microscopy (Bact., Hist.), indicator (pH 11.5-14.0)